Learn More
Bioworld BIOTIN SEPAROPOREAGAROSE)
Supplier:  Bioworld 201810613
Encompass
BIOTIN SEPAROPORE (AGAROSE) 6B-CL: Application: For the purification of avidin and streptavidin containing proteins. Specifications: Ligand: Biotin; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Cross-lin...
Catalog No. 50-197-821