Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Biotin-SLC | 1864003-57-5 | ≥97.0% | 250 MG
SDP

Catalog No. 5000341968
Encompass_Preferred

(+)-Biotin-SLC is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands (E3 ubiquitin ligase and target protein) connected by a linker
  • Exploits intracellular ubiquitin-proteasome system for target protein degradation
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month) under nitrogen

Catalog No. 50-003-41968 Supplier Medchemexpress LLC Supplier No. 250MGHY130887
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Biotin-SLC is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs.

  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands (E3 ubiquitin ligase and target protein) connected by a linker
  • Exploits intracellular ubiquitin-proteasome system for target protein degradation
  • For research use only
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month) under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.