Learn More
Medchemexpress LLC (+)-Biotin-SLC | 1864003-57-5 | ≥97.0% | 500 MG
SDP
Catalog No. 5000338552
Encompass_Preferred
(+)-Biotin-SLC is an alkyl chain-based PROTAC linker. It can be used in the synthesis of PROTACs, which are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
  • For research use only
Catalog No. 50-003-38552 Supplier Medchemexpress LLC Supplier No. HY130887500MG
(+)-Biotin-SLC is an alkyl chain-based PROTAC linker. It can be used in the synthesis of PROTACs, which are designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
  • For research use only