Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVIDITY LLC Biotin Solution, 500 μM d-biotin, 1.5 mL

Catalog No. NC9368711
Encompass

Avidity LLC's BIO200 Biotin Solution 0.5MM is a high-quality d-biotin solution designed for supplementing BirA biotin-protein ligase reactions. This product provides an essential component for in vitro biotinylation, ensuring optimal reaction conditions, especially when additional biotin is required.

  • Product type: Biotin solution
  • Concentration: 500 μM d-biotin
  • Volume: 1.5 mL
  • Buffer: 10 mM Bicine
  • pH: 8.3
  • Application: BirA biotin-protein ligase reactions and Supplement for in vitro biotin ligase reactions
  • Storage condition: Store at -20°C upon receipt
  • Shipping condition: Shipped at room temperature
  • Format: Liquid
  • Included in kits: BirA-500 and Bulk BirA in-vitro biotinylation reaction kits

Catalog No. NC9368711 Supplier AVIDITY LLC Supplier No. BIO200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avidity LLC's BIO200 Biotin Solution 0.5MM is a high-quality d-biotin solution designed for supplementing BirA biotin-protein ligase reactions. This product provides an essential component for in vitro biotinylation, ensuring optimal reaction conditions, especially when additional biotin is required.

  • Product type: Biotin solution
  • Concentration: 500 μM d-biotin
  • Volume: 1.5 mL
  • Buffer: 10 mM Bicine
  • pH: 8.3
  • Application: BirA biotin-protein ligase reactions and Supplement for in vitro biotin ligase reactions
  • Storage condition: Store at -20°C upon receipt
  • Shipping condition: Shipped at room temperature
  • Format: Liquid
  • Included in kits: BirA-500 and Bulk BirA in-vitro biotinylation reaction kits

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.