Learn More
IBA LifeSciences Biotin stock solution; 100 mM
Catalog No. NC3566473
Encompass_Preferred
Biotin stock solution for preparing a dissociation buffer when using the Strep-tag technology for cells. Biotin addition causes the rapid dissociation of isolation reagents from the surface.
Catalog No. NC3566473 Supplier IBA LifeSciences Supplier No. 66325010
Biotin stock solution for preparing a dissociation buffer when using the Strep-tag technology for cells. Biotin addition causes the rapid dissociation of isolation reagents from the surface.