Learn More
Biotium Biotin Streptavidin Conjugates Fluorescent Proteins Nucleotides Other Conjugates Biotin and Streptavidin Conjugates CF488A 1/EA
Catalog No. 5000499759
Encompass_Preferred
Potential applications of Biotin-CF Dye include detection of biotin binding sites and the degree of biotinylation of proteins and for the measurement of avidin and streptavidin in crude biological samples
Catalog No. 50-004-99759 Supplier Biotium Supplier No. 80034
Add to Cart
Add to Cart
Potential applications of Biotin-CF Dye include detection of biotin binding sites and the degree of biotinylation of proteins and for the measurement of avidin and streptavidin in crude biological samples