Learn More
Medchemexpress LLC Biotin-Substance P | 87468-58-4 | 99.7% | 1574.90 | 5 MG
SDP
Catalog No. 5000423307
Encompass_Preferred
Biotin-Substance P is the biotin tagged Substance P. Substance P (Neurokinin P) is a neuropeptide, acting as a neurotransmitter and as a neuromodulator in the CNS. The endogenous receptor for substance P is neurokinin 1 receptor (NK1-receptor, NK1R).
  • Biotin-tagged substance P
  • Functions as a neuropeptide
  • Acts as a neurotransmitter and neuromodulator in the CNS
  • Targets the neurokinin 1 receptor (NK1R)
  • High purity of 99.7%
  • White to off-white solid appearance
  • Soluble in H2O (20 mg/mL)
Catalog No. 50-004-23307 Supplier Medchemexpress LLC Supplier No. HYP25465MG
Biotin-Substance P is the biotin tagged Substance P. Substance P (Neurokinin P) is a neuropeptide, acting as a neurotransmitter and as a neuromodulator in the CNS. The endogenous receptor for substance P is neurokinin 1 receptor (NK1-receptor, NK1R).
  • Biotin-tagged substance P
  • Functions as a neuropeptide
  • Acts as a neurotransmitter and neuromodulator in the CNS
  • Targets the neurokinin 1 receptor (NK1R)
  • High purity of 99.7%
  • White to off-white solid appearance
  • Soluble in H2O (20 mg/mL)