Learn More
Cayman Chemical Biotin sulfone, Purity: ≥95%, CAS: 40720-05-6, 100 mg
SDP
Catalog No. 502946993
Encompass_Preferred
An oxidized form of biotin; binds to avidin with a binding ratio of 0.332 compared with biotin; has been used to immobilize oligosaccharides on avidin-coated surfaces for the detection of anti-C. albicans antibodies
Catalog No. 50-294-6993 Supplier Cayman Chemical Supplier No. 34144100 MG
An oxidized form of biotin; binds to avidin with a binding ratio of 0.332 compared with biotin; has been used to immobilize oligosaccharides on avidin-coated surfaces for the detection of anti-C. albicans antibodies