Learn More
AdipoGen Biotin Sulfone
SDP
Catalog No. 502561581
Encompass_Preferred
Chemical. CAS 40720-05-6. Formula C10H16N2O5S. Molecular Weight 276.31. Biotin sulfone is an oxidized form of biotin and is as efficient as biotin in streptavidin binding and it binds to avidin with a binding ratio of 0.332 compared with biotin. Biotinylated synthetic oligosaccharides are an attractive tool, as streptavidin coated plates and beads are commercially available and widely use in serological assays. As biotin is chemically reactive, and not compatible with carbohydrate protecting group manipulation, the use of biotin sulfone tagged oligomannosides BSTO for antigen immobilization in candidiasis or allergy diagnosis has been used. The use of biotin sulfone also offers a convenient solution for both oligosaccharide synthesis and immobilization on microspheres and surface plasmon resonance sensors.
Catalog No. 50-256-1581 Supplier AdipoGen Supplier No. CDXB0606M250
Chemical. CAS 40720-05-6. Formula C10H16N2O5S. Molecular Weight 276.31. Biotin sulfone is an oxidized form of biotin and is as efficient as biotin in streptavidin binding and it binds to avidin with a binding ratio of 0.332 compared with biotin. Biotinylated synthetic oligosaccharides are an attractive tool, as streptavidin coated plates and beads are commercially available and widely use in serological assays. As biotin is chemically reactive, and not compatible with carbohydrate protecting group manipulation, the use of biotin sulfone tagged oligomannosides BSTO for antigen immobilization in candidiasis or allergy diagnosis has been used. The use of biotin sulfone also offers a convenient solution for both oligosaccharide synthesis and immobilization on microspheres and surface plasmon resonance sensors.