Learn More
Medchemexpress LLC Biotin sulfone | 40720-05-6 | 99.8% | 276.31 | 10 MG
SDP
Catalog No. 5000436971
Encompass_Preferred
Biotin sulfone is a structural analog and metabolite of Biotin. It competitively inhibits Biotin in *Lactobacillus arabinosus* 17-5 and binds tightly to streptavidin.
  • Competitively inhibits biotin utilization by *Lactobacillus arabinosus*
  • Binds tightly to streptavidin
  • Soluble in DMSO at 250 mg/mL
  • Available as a solid
Catalog No. 50-004-36971 Supplier Medchemexpress LLC Supplier No. HY11326810MG
Biotin sulfone is a structural analog and metabolite of Biotin. It competitively inhibits Biotin in *Lactobacillus arabinosus* 17-5 and binds tightly to streptavidin.
  • Competitively inhibits biotin utilization by *Lactobacillus arabinosus*
  • Binds tightly to streptavidin
  • Soluble in DMSO at 250 mg/mL
  • Available as a solid