Learn More
Medchemexpress LLC Biotin sulfone | 40720-05-6 | 99.8% | 276.31 | 25 MG
SDP
Catalog No. 5000439282
Encompass_Preferred
Biotin sulfone is a structural analog and metabolite of Biotin. It competitively inhibits Biotin in Lactobacillus arabinosus 17-5. The inhibitory potency is regulated by medium additives, with the inhibition index increasing with aspartic acid, Tween 80, and a combination of lysine, threonine, and uracil. Biotin sulfone binds tightly to streptavidin with performance comparable to that of Biotin.
  • Structural analog and metabolite of Biotin
  • Competitively inhibits Biotin in Lactobacillus arabinosus 17-5
  • Binds tightly to streptavidin
  • Appearance: Solid, white to off-white
  • Can be stored for extended periods in powder form
  • Detailed solubility and dissolution protocols provided for in vitro and in vivo experiments
Catalog No. 50-004-39282 Supplier Medchemexpress LLC Supplier No. 25MGHY113268
Biotin sulfone is a structural analog and metabolite of Biotin. It competitively inhibits Biotin in Lactobacillus arabinosus 17-5. The inhibitory potency is regulated by medium additives, with the inhibition index increasing with aspartic acid, Tween 80, and a combination of lysine, threonine, and uracil. Biotin sulfone binds tightly to streptavidin with performance comparable to that of Biotin.
  • Structural analog and metabolite of Biotin
  • Competitively inhibits Biotin in Lactobacillus arabinosus 17-5
  • Binds tightly to streptavidin
  • Appearance: Solid, white to off-white
  • Can be stored for extended periods in powder form
  • Detailed solubility and dissolution protocols provided for in vitro and in vivo experiments