Learn More
Cayman Chemical Biotin Sulfoxide, Purity: ≥95%, CAS: 23015-53-4, 10 mg
SDP
Catalog No. 502942812
Encompass_Preferred
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide
Catalog No. 50-294-2812 Supplier Cayman Chemical Supplier No. 4017210 MG
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide