Learn More
Cayman Chemical Biotin Sulfoxide, Purity: ≥95%, CAS: 23015-53-4, 25 mg
SDP
Catalog No. 502942834
Encompass_Preferred
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide
Catalog No. 50-294-2834 Supplier Cayman Chemical Supplier No. 4017225 MG
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide