Learn More
Medchemexpress LLC Biotin-TEG-ATFBA | 1264662-85-2 | C27H37F4N7O6S | 25 MG
SDP
Catalog No. 5000374268
Encompass_Preferred
Biotin-TEG-ATFBA is a click chemistry reagent that incorporates a perfluorophenylazide group. This reagent forms a highly stable azene intermediate which undergoes insertion and addition reactions (non-intermolecular rearrangement) in moderate to good yields after photolysis. It also has an azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. Additionally, it can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. It is intended for research use only.
  • Contains a perfluorophenylazide group
  • Forms a highly stable azene intermediate upon photolysis
  • Undergoes insertion and addition reactions
  • Features an azide group for CuAAc with alkyne-containing molecules
  • Can undergo SPAAC with DBCO or BCN groups
Catalog No. 50-003-74268 Supplier Medchemexpress LLC Supplier No. 25MGHY151672
Biotin-TEG-ATFBA is a click chemistry reagent that incorporates a perfluorophenylazide group. This reagent forms a highly stable azene intermediate which undergoes insertion and addition reactions (non-intermolecular rearrangement) in moderate to good yields after photolysis. It also has an azide group, allowing it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. Additionally, it can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. It is intended for research use only.
  • Contains a perfluorophenylazide group
  • Forms a highly stable azene intermediate upon photolysis
  • Undergoes insertion and addition reactions
  • Features an azide group for CuAAc with alkyne-containing molecules
  • Can undergo SPAAC with DBCO or BCN groups