Learn More
Medchemexpress LLC Biotin-thalidomide | 2230857-87-9 | MFCD32851674 | 99.7% | 945.09 g/mol | C44H64N8O13S | 5 MG
SDP
Catalog No. 5000221271
Encompass_Preferred
Biotin-Thalidomide is a biotin-conjugated thalidomide derivative that binds the cereblon (CRBN) E3 ligase and is supplied for research use in target engagement studies and PROTAC synthesis. It is provided as a high-purity chemical probe for biochemical assays, affinity capture, and conjugation workflows; not for human therapeutic use.
  • High purity (≈99.7%) suitable for research applications
  • Cereblon-binding probe useful for PROTAC development
  • Biotin tag enables affinity capture and pull-down assays
  • Compatible with conjugation and synthetic workflows
  • Intended for research use only
Catalog No. 50-002-21271 Supplier Medchemexpress LLC Supplier No. HY1451525MG
Biotin-Thalidomide is a biotin-conjugated thalidomide derivative that binds the cereblon (CRBN) E3 ligase and is supplied for research use in target engagement studies and PROTAC synthesis. It is provided as a high-purity chemical probe for biochemical assays, affinity capture, and conjugation workflows; not for human therapeutic use.
  • High purity (≈99.7%) suitable for research applications
  • Cereblon-binding probe useful for PROTAC development
  • Biotin tag enables affinity capture and pull-down assays
  • Compatible with conjugation and synthetic workflows
  • Intended for research use only