Learn More
Medchemexpress LLC Biotin-thalidomide | 2230857-87-9 | MFCD32851674 | 99.7% | C44H64N8O13S | 10MG
SDP
Catalog No. 5000261865
Encompass_Preferred
Biotin-thalidomide is a biotin-conjugated thalidomide derivative that binds the cereblon (CRBN) E3 ligase and is supplied as a high-purity reagent for use in PROTAC synthesis and biochemical assays.
  • Binds cereblon (CRBN) E3 ligase
  • Suitable for PROTAC synthesis and biochemical binding studies
  • Facilitates affinity capture and pull-down assays via biotin tag
  • Provided as a high-purity reagent for research use
  • Characterized by molecular formula and SMILES for cheminformatics
Catalog No. 50-002-61865 Supplier Medchemexpress LLC Supplier No. HY14515210MG
Biotin-thalidomide is a biotin-conjugated thalidomide derivative that binds the cereblon (CRBN) E3 ligase and is supplied as a high-purity reagent for use in PROTAC synthesis and biochemical assays.
  • Binds cereblon (CRBN) E3 ligase
  • Suitable for PROTAC synthesis and biochemical binding studies
  • Facilitates affinity capture and pull-down assays via biotin tag
  • Provided as a high-purity reagent for research use
  • Characterized by molecular formula and SMILES for cheminformatics