Learn More
CPC Scientific Biotin-Tyr-Val-Ala-Asp-CMK 1MG
SDP
Catalog No. 502108960
Encompass
Biotinylated analog of the caspase-1 inhibitor.ONE-LETTER SEQUENCE: Biotin-YVAD-CMKMOLECULAR FORMULA: C32H45Cl1N6O9S1MOLECULAR WEIGHT:725.27STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [402474-75-3], SYNONYMS: Biotinyl-Tyr-Val-Ala-Asp-chloromethylketoneRESEARCH AREA: ApoptosisREFERENCES: Y.A. Lazebnik et al., Nature, 371, 346 (1994)
Catalog No. 50-210-8960 Supplier CPC Scientific Supplier No. CASP021B
Biotinylated analog of the caspase-1 inhibitor.ONE-LETTER SEQUENCE: Biotin-YVAD-CMKMOLECULAR FORMULA: C32H45Cl1N6O9S1MOLECULAR WEIGHT:725.27STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [402474-75-3], SYNONYMS: Biotinyl-Tyr-Val-Ala-Asp-chloromethylketoneRESEARCH AREA: ApoptosisREFERENCES: Y.A. Lazebnik et al., Nature, 371, 346 (1994)