Learn More
Apexbio Technology LLC Biotin-tyramide, 100mg. Cas: 41994-02-9 MFCD: MFCD27953026. Reagent used for tyramide signal amplification (TSA)
SDP
Supplier:  Apexbio Technology LLC A8011100
Encompass_Preferred
Biotin-tyramide is used for tyramide signal amplification (TSA) which is a powerful, patented technology that significantly enhances both chromogenic and fluorescent signals. TSA is easily integrated into standard nonradioactive in situ hybridization (ISH) or IHC protocols Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4983