Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biotin-Ubiquitin

Catalog No. PV4380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4380 Supplier Invitrogen™ Supplier No. PV4380
Add to Cart
Add to Cart

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination.

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (Cat. No. PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Conjugate Biotin
Content And Storage Biotin labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340/490, 520 nm
For Use With (Equipment) Microplate Reader
Label or Dye Biotin
Label Type Biotin & Other Haptens
Readout End Point
Shipping Condition Dry Ice
Substrate Properties Protein-Based Substrate
Technique TR-FRET
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.