Learn More
Cayman Chemical Biotin-VAD-FMK, Purity: ≥95%, CAS: 1135688-15-1, 1 mg
SDP
Catalog No. 502936461
Encompass_Preferred
A biotin-conjugated caspase inhibitor; an affinity probe that allows for detection and immobilization of caspases using the biotin ligand; inhibits caspase-3-like activity in a dose-dependent manner in hepatocytes stimulated with TNF-α and ActD; binds to caspase-3 and caspase-8 in hepatocyte lysates
Catalog No. 50-293-6461 Supplier Cayman Chemical Supplier No. 243051 MG
A biotin-conjugated caspase inhibitor; an affinity probe that allows for detection and immobilization of caspases using the biotin ligand; inhibits caspase-3-like activity in a dose-dependent manner in hepatocytes stimulated with TNF-α and ActD; binds to caspase-3 and caspase-8 in hepatocyte lysates