Learn More
Medchemexpress LLC HY-100894 1mg Medchemexpress, Biotin-VAD-FMK CAS:1135688-15-1 Purity:>98%
SDP
Catalog No. 501967788
Encompass_Preferred
Medchemexpress, HY-100894 1mg Biotin-VAD-FMK CAS:1135688-15-1 Biotin-VAD-FMK is a cell permeable, irreversible biotin-labeled caspase inhibitor, used to identify active caspases in cell lysates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7788 Supplier Medchemexpress LLC Supplier No. HY1008941MG
Medchemexpress, HY-100894 1mg Biotin-VAD-FMK CAS:1135688-15-1 Biotin-VAD-FMK is a cell permeable, irreversible biotin-labeled caspase inhibitor, used to identify active caspases in cell lysates. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.