Learn More
Medchemexpress LLC Biotin-val-ala-asp(ome)-fluoromethyl ketone | 1135688-15-1 | >98.0% | 672.81 g/mol | C30H49FN6O8S | 100 MG
SDP
Catalog No. 5000211791
Encompass_Preferred
Biotin-VAD-FMK is a cell-permeable, irreversible, biotin-labeled caspase inhibitor used to identify active caspases in cell lysates by covalently trapping active enzymes and enabling detection or affinity enrichment via the biotin tag.
  • Cell-permeable, irreversible caspase inhibitor for activity-based labeling.
  • Biotin tag enables detection and streptavidin-based enrichment.
  • Suitable for profiling active caspases in cell lysates and biochemical assays.
  • Available as a solid white to light yellow powder for convenient handling.
  • High purity (>98.0%) for reliable experimental results.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
Catalog No. 50-002-11791 Supplier Medchemexpress LLC Supplier No. HY100894100MG
Biotin-VAD-FMK is a cell-permeable, irreversible, biotin-labeled caspase inhibitor used to identify active caspases in cell lysates by covalently trapping active enzymes and enabling detection or affinity enrichment via the biotin tag.
  • Cell-permeable, irreversible caspase inhibitor for activity-based labeling.
  • Biotin tag enables detection and streptavidin-based enrichment.
  • Suitable for profiling active caspases in cell lysates and biochemical assays.
  • Available as a solid white to light yellow powder for convenient handling.
  • High purity (>98.0%) for reliable experimental results.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).