Learn More
Medchemexpress LLC Biotin-Val-Ala-Asp(OMe)-FMK | 1135688-15-1 | ≥95.0% | 672.81 g/mol | C30H49FN6O8S | 50 MG
SDP
Catalog No. 5000212382
Encompass_Preferred
Biotin-VAD-FMK is a biotin-labeled, cell-permeable, irreversible pan-caspase inhibitor used to identify and label active caspases in cell lysates. It serves as an affinity probe for detection or capture of active caspases in apoptosis research and protease-profiling workflows, and is compatible with biochemical assays and pull-down experiments.
  • Biotin-labeled for affinity capture and detection.
  • Cell-permeable, irreversible caspase inhibitor.
  • Enables identification and labeling of active caspases in cell lysates.
  • Compatible with biochemical assays, protease profiling, and pull-down workflows.
  • Supplied as a solid with defined molecular weight and formula for experimental planning.
Catalog No. 50-002-12382 Supplier Medchemexpress LLC Supplier No. HY10089450MG
Biotin-VAD-FMK is a biotin-labeled, cell-permeable, irreversible pan-caspase inhibitor used to identify and label active caspases in cell lysates. It serves as an affinity probe for detection or capture of active caspases in apoptosis research and protease-profiling workflows, and is compatible with biochemical assays and pull-down experiments.
  • Biotin-labeled for affinity capture and detection.
  • Cell-permeable, irreversible caspase inhibitor.
  • Enables identification and labeling of active caspases in cell lysates.
  • Compatible with biochemical assays, protease profiling, and pull-down workflows.
  • Supplied as a solid with defined molecular weight and formula for experimental planning.