Learn More
Medchemexpress LLC Biotin-vitamin B12 | 98.4% | 1581.66 | C73H102CoN16O16PS | 10 MG
SDP
Catalog No. 5000298890
Encompass_Preferred
Biotin-Vitamin B12 is a biotinylated form of cyanocobalamin (vitamin B12) used as a biochemical reagent and affinity probe. The biotin tag enables capture or detection with streptavidin/avidin systems. Supplied as a solid with high purity, it is formulated for in vitro use and can be prepared for in vivo dosing following manufacturer guidance.
Catalog No. 50-002-98890 Supplier Medchemexpress LLC Supplier No. HYB0315A10MG
Biotin-Vitamin B12 is a biotinylated form of cyanocobalamin (vitamin B12) used as a biochemical reagent and affinity probe. The biotin tag enables capture or detection with streptavidin/avidin systems. Supplied as a solid with high purity, it is formulated for in vitro use and can be prepared for in vivo dosing following manufacturer guidance.