Learn More
Seracare Life Sciences Inc BIOTIN WASH KIT
Supplier:  Seracare Life Sciences Inc 59600015
Encompass_Preferred
10X Wash solution concentrate specially formulated for use with biotin-streptavidin systems. Recommended for use in biotin-streptavidin applications to reduce background due to nonspecific binding of enzyme-labeled streptavidin to nylon membranes an in tissues. Applications for Biotin Wash Solution include nucleic acid blotting, colony and plaque lifts, Western blotting, and in situ hybridization.
Catalog No. 50-674-12