Learn More
Cayman Chemical Biotin-X-NHS, Purity: ≥95%, CAS: 72040-63-2, 100 mg
SDP
Catalog No. 502953738
Encompass_Preferred
A compound used to attach biotin to primary amines, such as those found on lysines on the surface of proteins; contains six-atom spacer arm
Catalog No. 50-295-3738 Supplier Cayman Chemical Supplier No. 13316100 MG
A compound used to attach biotin to primary amines, such as those found on lysines on the surface of proteins; contains six-atom spacer arm