Learn More
Biotium Biotin-X NTA (OR Biotin-X NITRILOTRIACET
Catalog No. 501964811
Encompass_Preferred
Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt) is used to detect histidine-tagged proteins (as little as 0.1 pmol) immobilized on nitrocellulose membranes. Biotin-X NTA can be removed from the histidine-tagged protein at pH 4.8, allowing the blot to be reanalyzed with another probe. Properties: White solid soluble in water; Store at 4°C; C26H40K3N5O9S; MW: 715.98
Catalog No. 50-196-4811 Supplier Biotium Supplier No. 90056
Add to Cart
Edge
Add to Cart
Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt) is used to detect histidine-tagged proteins (as little as 0.1 pmol) immobilized on nitrocellulose membranes. Biotin-X NTA can be removed from the histidine-tagged protein at pH 4.8, allowing the blot to be reanalyzed with another probe. Properties: White solid soluble in water; Store at 4°C; C26H40K3N5O9S; MW: 715.98