Learn More
Invitrogen™ Biotin-X, SE (6-((Biotinoyl)Amino)Hexanoic Acid, Succinimidyl Ester (Biotinamidocaproate, N-Hydroxysuccinimidyl Ester))
Description
The amine-reactive biotin-X, SE can be used to attach this important hapten to biomolecules of interest for subsequent detection with streptavidin, avidin or NeutrAvidin™ biotin-binding protein.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Biotin-XX SE |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Biotin-X |
| Label Type | Biotin and Other Haptens |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.