Learn More
AdipoGen Biotin-XX
SDP
Catalog No. 502561575
Encompass_Preferred
Chemical. CAS 89889-51-0. Formula C22H38N4O5S. Molecular Weight 470.63. Biotinylated cross-linking reagent. Biotin-XX has a 14 atom spacer designated by XX to facilitate reactivity with the molecule and eventual accessibility of the biotin to avidin or streptavidin.
Catalog No. 50-256-1575 Supplier AdipoGen Supplier No. CDXB0632M100
Chemical. CAS 89889-51-0. Formula C22H38N4O5S. Molecular Weight 470.63. Biotinylated cross-linking reagent. Biotin-XX has a 14 atom spacer designated by XX to facilitate reactivity with the molecule and eventual accessibility of the biotin to avidin or streptavidin.