Learn More
Cayman Chemical BIotIn-XX hydrazIde 10mg
SDP
Catalog No. 502952539
Encompass_Preferred
A biotinylating reagent that contains two aminohexanoic spacers to increase the efficiency of avidin-binding by alleviating steric hindrance
Catalog No. 50-295-2539 Supplier Cayman Chemical Supplier No. 1330410 MG
A biotinylating reagent that contains two aminohexanoic spacers to increase the efficiency of avidin-binding by alleviating steric hindrance