Learn More
Medchemexpress LLC Biotin-XX hydrazide | 211237-33-1 | 95.4% | 484.66 g/mol | C22H40N6O4S | 5MG
SDP
Supplier:  Medchemexpress LLC HY1255015MG
Encompass_Preferred
Biotin-XX hydrazide is a carbonyl-reactive biotinylation reagent that contains two aminohexanoic acid spacers to improve avidin-binding efficiency. It couples to carbonyl groups on biomolecules to produce biotinylated targets suitable for detection, enrichment, and affinity capture using streptavidin or avidin methods.
  • Contains two aminohexanoic acid spacers to enhance binding efficiency.
  • Reacts selectively with carbonyl groups for targeted labeling.
  • Enables affinity capture and detection with streptavidin or avidin systems.
  • Supplied as a 5 mg package suitable for laboratory-scale experiments.
  • High purity as determined by RP-HPLC for reliable results.
Catalog No. 50-002-36290