Learn More
Cayman Chemical Biotin-XX hydrazide, Purity: ≥95%, CAS: 211237-33-1, 5 mg
SDP
Catalog No. 502952445
Encompass_Preferred
A biotinylating reagent that contains two aminohexanoic spacers to increase the efficiency of avidin-binding by alleviating steric hindrance
Catalog No. 50-295-2445 Supplier Cayman Chemical Supplier No. 133045 MG
A biotinylating reagent that contains two aminohexanoic spacers to increase the efficiency of avidin-binding by alleviating steric hindrance