Learn More
Medchemexpress LLC Biotinamidocaproyl hydrazide | 109276-34-8 | MFCD00077659 | 99.8% | 371.50 g/mol | C16H29N5O3S | 10 MG
SDP
Catalog No. 5000248926
Encompass_Preferred
Biotin LC hydrazide is a long-chain biotinylation reagent used to derivatize periodate-oxidized carbohydrates on glycoproteins and other biomolecules. The hydrazide moiety reacts with aldehydes generated by mild oxidation to form stable hydrazone linkages, while the long-chain spacer arm improves accessibility for detection and affinity capture. Intended for research use only.
  • Reacts with periodate-oxidized carbohydrates and glycoproteins.
  • Introduces a biotin tag via hydrazide chemistry for avidin/streptavidin detection.
  • Long-chain spacer reduces steric hindrance and improves binding accessibility.
  • Suitable for labeling, detection, and affinity purification workflows.
  • Provided as a stable white solid for easy handling and storage.
Catalog No. 50-002-48926 Supplier Medchemexpress LLC Supplier No. HY10188510MG
Biotin LC hydrazide is a long-chain biotinylation reagent used to derivatize periodate-oxidized carbohydrates on glycoproteins and other biomolecules. The hydrazide moiety reacts with aldehydes generated by mild oxidation to form stable hydrazone linkages, while the long-chain spacer arm improves accessibility for detection and affinity capture. Intended for research use only.
  • Reacts with periodate-oxidized carbohydrates and glycoproteins.
  • Introduces a biotin tag via hydrazide chemistry for avidin/streptavidin detection.
  • Long-chain spacer reduces steric hindrance and improves binding accessibility.
  • Suitable for labeling, detection, and affinity purification workflows.
  • Provided as a stable white solid for easy handling and storage.