Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Biotinamidoethyldisulfanyl(2-Aminoethane) Trifluoroacetic Acid

Catalog No. 91366950MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-366-950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-366-950MG Supplier Sigma Aldrich Supplier No. 91366950MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biotinamidoethyldisulfanyl(2-aminoethane) trifluoroacetic acid (Biotin-SS-NH2 TFA) is a cleavable, biotinylated crosslinkers. The linker can also be used for synthesis of chemical biology tools for labeling target proteins in biological experiments and assays. It possesses dual functionality: enrichment via the biotin and an embedded disulfide bridge for cleavage under reducing conditions.Fluorometric assay for tissue transglutaminase-mediated transamidation activityConvenient Synthesis of Photoaffinity Probes and Evaluation of Their Labeling AbilitiesA simple photo-affinity labeling protocol

Specifications

Molecular Formula C20H25N3O2
Linear Formula C20H25N3O2
Quantity 50 mg
Synonym N-((1R,2R)-2-(Benzyloxy)cyclopentyl)-3-(3-(but-3-yn-1-yl)-3H-diazirin-3-yl)propanamide; Fully functionalized enantioprobe; Ligandability probe
Molecular Weight (g/mol) 339.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.