Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Biotinamidohexanoic Acid Hydrazide

Catalog No. B377025MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B377025MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B377025MG Supplier Sigma Aldrich Supplier No. B377025MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biotinylation reagent with aminocaproic spacer for aldehyde groups (e.g., periodate-oxidized sugars) and carboxylic acids. Typically used for coupling to glycoproteins through the carbohydrate by hydrazone bond formation.

Specifications

CAS 5460-29-7
Melting Point 72°C to 74°C
Molecular Formula C11H10BrNO2
Linear Formula C11H10BrNO2
MDL Number MFCD00005904
Quantity 100 g
Molecular Weight (g/mol) 268.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.