Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Biotinamidohexanoyl-6-Aminohexanoic Acid N-Hydroxysuccinimide Ester

Catalog No. B329550MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
30 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B329550MG 30 mg
B329510MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B329550MG Supplier Sigma Aldrich Supplier No. B329550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biotinylation reagent incorporating an extended 14-atom spacer can reduce steric hindrance in binding avidin or streptavidin to some biotinylated compounds and increase sensitivity for detection. Typically coupled to primary amine in the pH range 6.5-8.5.

Specifications

CAS 92-52-4
Melting Point 68°C to 70°C
Boiling Point 255°C
Molecular Formula C12H10
Linear Formula C6H5C6H5
MDL Number MFCD00003054
Quantity 30 mg
Molecular Weight (g/mol) 154.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.