Learn More
Cayman Chemical Biotin-H10 (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502915345
Encompass_Preferred
A biotinylated form of H10; has been used in ELISA and co-immunoprecipitation assays to assess AGR2 levels in cancer cells
Catalog No. 50-291-5345 Supplier Cayman Chemical Supplier No. 3700510 MG
A biotinylated form of H10; has been used in ELISA and co-immunoprecipitation assays to assess AGR2 levels in cancer cells