Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human DR3 (TNFRSF25) Protein, Fc-Avitag, 25 μg (HEK293, MALS verified)

Catalog No. NC3385711
Encompass_Preferred

Biotinylated human DR3 (TNFRSF25) extracellular domain fused to a human IgG1 Fc and a biotin acceptor tag, expressed in HEK293 cells and verified by SEC-MALS. Supplied lyophilized (25 μg), this reagent is intended for binding studies and assay development, including ELISA and SPR.

  • Expressed in HEK293 cells.
  • Contains amino acids Gln 25-Gln 199 of human DR3 (UniProt Q93038-1).
  • Carries a C-terminal human IgG1 Fc and a biotin acceptor tag; enzymatically biotinylated.
  • Calculated molecular weight 47.2 kDa; migrates 55-65 kDa on SDS-PAGE.
  • Purity greater than 90% by SDS-PAGE and greater than 95% by SEC-MALS.
  • Lyophilized format, 25 μg; formulated in PBS with trehalose.
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw.

Catalog No. NC3385711 Supplier ACROBiosystems Supplier No. DR3H82F325UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated human DR3 (TNFRSF25) extracellular domain fused to a human IgG1 Fc and a biotin acceptor tag, expressed in HEK293 cells and verified by SEC-MALS. Supplied lyophilized (25 μg), this reagent is intended for binding studies and assay development, including ELISA and SPR.

  • Expressed in HEK293 cells.
  • Contains amino acids Gln 25-Gln 199 of human DR3 (UniProt Q93038-1).
  • Carries a C-terminal human IgG1 Fc and a biotin acceptor tag; enzymatically biotinylated.
  • Calculated molecular weight 47.2 kDa; migrates 55-65 kDa on SDS-PAGE.
  • Purity greater than 90% by SDS-PAGE and greater than 95% by SEC-MALS.
  • Lyophilized format, 25 μg; formulated in PBS with trehalose.
  • Store lyophilized at -20°C or lower; avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.