Learn More
Cayman Chemical BIotInS-sulfoxIde 50mg
SDP
Catalog No. 502947146
Encompass_Preferred
An inactive metabolite of biotin; has been found in E. coli, where it is used as a source of biotin
Catalog No. 50-294-7146 Supplier Cayman Chemical Supplier No. 3414550 MG
An inactive metabolite of biotin; has been found in E. coli, where it is used as a source of biotin