Learn More
Cayman Chemical BIotInS-sulfoxIde 5mg
SDP
Catalog No. 502946657
Encompass_Preferred
An inactive metabolite of biotin; has been found in E. coli, where it is used as a source of biotin
Catalog No. 50-294-6657 Supplier Cayman Chemical Supplier No. 341455 MG
An inactive metabolite of biotin; has been found in E. coli, where it is used as a source of biotin