Learn More
Bachem Biotinyl-Amylin (mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4089772.0500
Encompass_Preferred
Biotinyl-Amylin (mouse, rat) Trifluoroacetate, Biotinyl-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-Arg-Ser-Ser-Asn-Asn-Leu-Gly-Pro-Val-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH2, 1678414-88-4, C177H286N54O55S3, Mr 4146.75, 0.5mg.
Catalog No. 50-194-6693