Learn More
Medchemexpress LLC Biotinyl-Amylin (mouse, rat) TFA | 1678414-88-4 | >95% | 4146.69 | 1 MG
SDP
Catalog No. 5000399208
Encompass_Preferred
Biotinyl-Amylin (mouse, rat) TFA is a biotinylated form of Amylin (mouse, rat), a 37-amino acid pancreatic protein. This compound plays a role in controlling various metabolic events, including glycemia and lacticemia. It is designed for laboratory use.
  • Biotinylated derivative of amylin (mouse, rat)
  • Involved in the control of metabolic events
  • 37-amino acid pancreatic protein
  • Used in research related to carboxylic acids and pharmaceutical standards
  • Categorized under enzyme, peptide, and protein related compounds
  • Intended for laboratory use only
Catalog No. 50-003-99208 Supplier Medchemexpress LLC Supplier No. HYP4928A1MG
Biotinyl-Amylin (mouse, rat) TFA is a biotinylated form of Amylin (mouse, rat), a 37-amino acid pancreatic protein. This compound plays a role in controlling various metabolic events, including glycemia and lacticemia. It is designed for laboratory use.
  • Biotinylated derivative of amylin (mouse, rat)
  • Involved in the control of metabolic events
  • 37-amino acid pancreatic protein
  • Used in research related to carboxylic acids and pharmaceutical standards
  • Categorized under enzyme, peptide, and protein related compounds
  • Intended for laboratory use only