Learn More
Enzo Life Sciences Biotinyl-DEVD-CHO (5mg)
SDP
Catalog No. 502008247
Encompass_Preferred
Reversible inhibitor of caspase-3 and -7. Useful for the purification or extract depletion of caspase-3 in conjunction with streptavidin conjugates. Alternative name: Caspase-3 inhibitor (aldehyde), Caspase-7 inhibitor (aldehyde), CPP32 affinity reagent. Formula: C28H42N6O12S. MW: 686.7. Purity: ≥99% (HPLC). Appearance: White powder. Formulation: Lyophilized. Solubility: Soluble in 50% acetic acid with agitation (1mg/ml). Long Term Storage: -20°C. Handling: Packaged under inert gas.
Catalog No. 50-200-8247 Supplier Enzo Life Sciences Supplier No. ALX260034M005
Reversible inhibitor of caspase-3 and -7. Useful for the purification or extract depletion of caspase-3 in conjunction with streptavidin conjugates. Alternative name: Caspase-3 inhibitor (aldehyde), Caspase-7 inhibitor (aldehyde), CPP32 affinity reagent. Formula: C28H42N6O12S. MW: 686.7. Purity: ≥99% (HPLC). Appearance: White powder. Formulation: Lyophilized. Solubility: Soluble in 50% acetic acid with agitation (1mg/ml). Long Term Storage: -20°C. Handling: Packaged under inert gas.