Learn More
Bachem Biotinyl-eAhx-o-Conotoxin GVIA Trifluoroacetate
SDP
Supplier:  Bachem 4045700.1000
Encompass_Preferred
Biotinyl-eAhx-o-Conotoxin GVIA Trifluoroacetate, Biotinyl-eAhx-Cys-Lys-Ser-Hyp-Gly-Ser-Ser-Cys-Ser-Hyp-Thr-Ser-Tyr-Asn-Cys-Cys-Arg-Ser-Cys-Asn-Hyp-Tyr-Thr-Lys-Arg-Cys-Tyr-NH2, 151928-23-3, C136H207N41O46S7, Mr 3376.85, 1mg. This biotinylated o-conotoxin GVIA is a useful ligand for the characterization of calcium channels.
Catalog No. 50-260-034