Learn More
CPC Scientific Biotin-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GLUC001B
Encompass
ONE-LETTER SEQUENCE: Biotin-HSQGTFTSDYSKYLDSRRAQDFVQWLMNTMOLECULAR FORMULA: C163H239N45O51S2MOLECULAR WEIGHT:3709.10STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802086-28-7], SYNONYMS: Biotinyl-Glucagon (1-29) (human, rat, porcine)RESEARCH AREA: Diabetes
Catalog No. 50-210-9360