Learn More
Cayman Chemical Biotinyl hexylamine, Purity: ≥98%, CAS: 65953-56-2, 50 mg
SDP
Catalog No. 502978923
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of ANANAS and chemotherapeutic conjugates
Catalog No. 50-297-8923 Supplier Cayman Chemical Supplier No. 3122650 MG
A synthetic intermediate; has been used in the synthesis of ANANAS and chemotherapeutic conjugates