Learn More
CPC Scientific Biotinyl-Pro-His-Ala-Gln-Leu-Leu-Arg-Val-Gly-Cys-Val-Leu-Gly-Thr-Cys-Gln-Val-Gln-Asn-Leu-Ser-His-Arg-Leu-Trp-Gln-Leu-Val-Arg-Pro-Ser-Gly-Arg-Arg-Asp-Ser-Ala-Pro-Val-Asp-Pro-Ser-Ser-Pro-His-Ser-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HITM003A
Encompass
ONE-LETTER SEQUENCE: Biotinyl-PHAQLLRVGCVLGTCQVQNLSHRLWQLVRPSGRRDSAPVDPSSPHSY-NH2MOLECULAR FORMULA: C236H375N77O66S3MOLECULAR WEIGHT:5443.25STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-210-9531