Learn More
CPC Scientific Biotinyl-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LL37004A
Encompass
SEQUENCE: Biotinyl-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Biotinyl-LLGDFFRKSKEKIGKEFKRIVQRIKDFLRNLVPRTES-NH2MOLECULAR FORMULA: C215H355N63O54S1MOLECULAR WEIGHT: 4718.64STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:
Tsai, Pei-Wen, et al. PloS One 6.3 (2011): e17755.
Srakaew, Nopparat, et al. Human Reproduction 29.4 (2014): 683-696.SKU(s): LL37-004A, LL37-004B
Catalog No. 50-210-9687