Learn More
Medchemexpress LLC Biotinyl-NH-PEG3-C3-amido-C3-COOH | 1202761-21-4 | 99.3% | 100 MG
SDP
Catalog No. 5000338668
Encompass_Preferred
Biotinyl-NH-PEG3-C3-amido-C3-COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This solid, white to off-white compound offers high purity and is intended for research applications.
  • High purity of 99.25%
  • PEG-based PROTAC linker
  • Suitable for synthesis of PROTACs
  • Soluble in DMSO and methanol
  • For research use only
Catalog No. 50-003-38668 Supplier Medchemexpress LLC Supplier No. HY130142A100MG
Biotinyl-NH-PEG3-C3-amido-C3-COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This solid, white to off-white compound offers high purity and is intended for research applications.
  • High purity of 99.25%
  • PEG-based PROTAC linker
  • Suitable for synthesis of PROTACs
  • Soluble in DMSO and methanol
  • For research use only