Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotinyl-NH-PEG3-C3-amido-C3-COOH | 1202761-21-4 | 99.3% | 50 MG
SDP

Catalog No. 5000338561
Encompass_Preferred

Biotinyl-NH-PEG3-C3-amido-C3-COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • Solid appearance
  • White to off-white color
  • Soluble in DMSO (250 mg/mL)
  • Soluble in methanol (≥ 100 mg/mL)
  • Molecular weight: 560.70
  • Molecular formula: C25H44N4O8S

Catalog No. 50-003-38561 Supplier Medchemexpress LLC Supplier No. HY130142A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinyl-NH-PEG3-C3-amido-C3-COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • Solid appearance
  • White to off-white color
  • Soluble in DMSO (250 mg/mL)
  • Soluble in methanol (≥ 100 mg/mL)
  • Molecular weight: 560.70
  • Molecular formula: C25H44N4O8S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.